missing translation for 'onlineSavingsMsg'
Learn More

Poly(methylhydrosiloxane), trimethylsilyl terminated

Product Code. 11966011 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
This item is not returnable. View return policy

Product Code. 11966011

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 63148-57-2 | (CH4OSi)n

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63148-57-2
Molecular Formula (CH4OSi)n
MDL Number MFCD00084478
InChI Key QNWOFLWXQGHSRH-UHFFFAOYSA-N
SMILES C[SiH](O[Si](C)(C)C)O[Si](C)(C)C

Specifications

Density 1.0060g/mL
Flash Point 204°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula CH3H9OSi-(CH4OSi)n-CH3H9Si
Quantity 100 mL
Specific Gravity 1.006
Chemical Name or Material Poly(methylhydrosiloxane)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.