missing translation for 'onlineSavingsMsg'
Learn More

Poly(N-vinylcarbazole), Secondary Standard, W.W. 135,6000, M.N. 56,400

Product Code. 10700985 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10700985 25 g 25g
10367431 5 g 5g
10624872 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10700985 Supplier Thermo Scientific Acros Supplier No. 329690250
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 25067-59-8 | (C14H11N)n | 193.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25067-59-8
Molecular Formula (C14H11N)n
Molecular Weight (g/mol) 193.25
MDL Number MFCD00134336
InChI Key KKFHAJHLJHVUDM-UHFFFAOYSA-N
Synonym 9-vinylcarbazole, n-vinylcarbazole, 9-vinyl-9h-carbazole, 9h-carbazole, 9-ethenyl, n-vinyl carbazole, vinylcarbazole, 1-vinylcarbazole, poly 9-vinylcarbazole, carbazole, 9-vinyl, poly-n-vinylcarbazole
PubChem CID 15143
SMILES *-CC(-*)N1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Color White
Physical Form Crystalline Powder
Density 1.2000g/mL
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in benzene, chloroform, dichloromethane,, dioxane, thf, toluene
Quantity 25 g
IUPAC Name 9-ethenylcarbazole
Infrared Spectrum Authentic
Loss on Drying 3% max.
Packaging Glass bottle
Refractive Index 1.683
Specific Gravity 1.2
Chemical Name or Material Poly(N-vinylcarbazole), Secondary standard
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.