missing translation for 'onlineSavingsMsg'
Learn More

Poly(N-vinylcarbazole), approx. M.W. 90,000

Product Code. 10225710 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10225710 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10225710 Supplier Thermo Scientific Acros Supplier No. 179030250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 25067-59-8 | (C14H11N)n | 193.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Color White
CAS 25067-59-8
Physical Form Crystalline Powder
Molecular Formula (C14H11N)n
MDL Number MFCD00134336
Density 1.2000g/mL
Synonym 9-vinylcarbazole, n-vinylcarbazole, 9-vinyl-9h-carbazole, 9h-carbazole, 9-ethenyl, n-vinyl carbazole, vinylcarbazole, 1-vinylcarbazole, poly 9-vinylcarbazole, carbazole, 9-vinyl, poly-n-vinylcarbazole
Quantity 25 g
InChI Key KKFHAJHLJHVUDM-UHFFFAOYSA-N
SMILES *-CC(-*)N1C2=C(C=CC=C2)C2=C1C=CC=C2
IUPAC Name 9-ethenyl-9H-carbazole
Molecular Weight (g/mol) 193.25
PubChem CID 15143
Infrared Spectrum Authentic
Loss on Drying 3% max.
Packaging Glass bottle
Chemical Name or Material Poly(N-vinylcarbazole), Secondary standard
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.