missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Poly-Net™ Close-mesh High-density Polyethylene Protective Netting

Product Code. 11786018 Shop All Cole-Parmer Products
Change view
Click to view available options
Material:
Polyethylene
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
11786018 Polyethylene Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11786018 Supplier Cole-Parmer™ Supplier No. 0940515
Big Savings for Better Separation
Use promo code 27992
View all promotions
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Provides protection for laboratory glassware

  • Fits securely over most small and large diameter lab vessels
  • Absorbs impact and reduces breakage
  • Safeguards against flying glass
  • This tough, high-density polyethylene (HDPE) resilient protective netting fits snugly over laboratory glassware to absorb impact, reduce breakage, and safeguard against flying glass
  • Simply cut to desired length to fit most vessels
  • Use smaller diameter sizes for instruments such as thermometers, pipettes, electrodes, tubes and syringes
  • Use larger diameter sizes for bottles, flasks, beakers, funnels and columns

Specifications

Type Poly-Net Protective Netting
Color Orange
Temperature (English) Max. 175°F
Temperature (Metric) Max 80°C
For Use With (Application) Provides Protection for Laboratory Glassware
Material Polyethylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.