missing translation for 'onlineSavingsMsg'
Learn More

Poly(phenylene sulfide), powder, nominal M.W. 10000, Thermo Scientific™

Product Code. 10732031 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
5g; Glass bottle
Product Code.
10732031 5g; Glass bottle
1 options
This item is not returnable. View return policy

Product Code. 10732031

Brand: Thermo Scientific Acros 178850050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Color Tan to White
CAS 9016-75-5
Physical Form Powder
Linear Formula (-C6H4S-)n
MDL Number MFCD00084445
Density 1.3600g/mL
Quantity 5 g
Infrared Spectrum Authentic
Loss on Drying 1% max.
Packaging Glass bottle
Specific Gravity 1.36
Chemical Name or Material Poly(phenylene sulfide), nominal M.W. 10000
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.