missing translation for 'onlineSavingsMsg'
Learn More

Poly(propylene glycol), average M.W. 425

Product Code. 10479890 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
250 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
2.5kg
250g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10479890 500 g Glass bottle 500g
10154400 5 g Glass bottle 5g
10338190 250 g Glass bottle 250g
11916981 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10479890 Supplier Thermo Scientific Acros Supplier No. 192155000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 25322-69-4 | (C3H6O)n | 134.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25322-69-4
Molecular Formula (C3H6O)n
Molecular Weight (g/mol) 134.18
MDL Number MFCD00084446
InChI Key DNIAPMSPPWPWGF-UHFFFAOYNA-N
Synonym polypropylene glycol, polyoxypropylene, 2-2-hydroxypropoxy propan-1-ol, emkapyl, lineartop e, niax ppg, polyoxypropylene glycol, methyloxirane homopolymer, desmophen 360c, 2-2-hydroxypropoxy-1-propanol
PubChem CID 32881
IUPAC Name 2-(2-hydroxypropoxy)propan-1-ol
SMILES [H]OC(C)CO

Specifications

Boiling Point 200.0°C
Melting Point -25.0°C
Linear Formula H[OCH(CH3)CH2]nOH
Density 1.0000g/mL
Solubility Information Solubility in water: completely soluble. Other solubilities: soluble in ethanol, acetone, benzene, toluene,, chloroform, dioxane, ether, thf
Quantity 500 g
Flash Point 185°C
Packaging Glass bottle
Refractive Index 1.4465 to 1.4505
Specific Gravity 1
Viscosity 80 mm2/s (25°C)
Chemical Name or Material Poly(propylene glycol), Average M.W. 425
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.