missing translation for 'onlineSavingsMsg'
Learn More

Poly(sodium-p-styrenesulfonate), average M.W. 70.000

Product Code. 10328550 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10328550 100 g Plastic bottle 100g
10012830 5 g Glass Bottle 5g
10144800 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10328550 Supplier Thermo Scientific Acros Supplier No. 222271000
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 25704-18-1 | (C8H7NaO3S)n | 184.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25704-18-1
Molecular Formula (C8H7NaO3S)n
Molecular Weight (g/mol) 184.21
MDL Number MFCD00084449
InChI Key MAGFQRLKWCCTQJ-UHFFFAOYSA-N
Synonym 4-styrenesulfonic acid sodium salt
PubChem CID 131675404
SMILES [Na]OS(=O)(=O)C1=CC=C(C=C1)C(-*)C-*

Specifications

Color Tan to White
Physical Form Powder
Quantity 100 g
Infrared Spectrum Authentic
Packaging Plastic bottle
Linear Formula [-CH2CH(C6H4SO3Na)-]n
Solubility Information Solubility in water: soluble. Other solubilities: soluble in lower glycerols
IUPAC Name 4-ethenylbenzenesulfonic acid;molecular hydrogen;sodium
Chemical Name or Material Poly(sodium-p-styrenesulfonate), Average M.W. 70.000

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.