missing translation for 'onlineSavingsMsg'
Learn More

Poly(styrene), average M.W. 260,000

Product Code. 10337790 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10337790 25 g Glass bottle 25g
10657744 100 g Glass bottle 100g
10636002 500 g Glass bottle 500g
10002380 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10337790 Supplier Thermo Scientific Acros Supplier No. 178890250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 9003-53-6 | (C8H8)n | 104.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9003-53-6
Molecular Formula (C8H8)n
Molecular Weight (g/mol) 104.15
MDL Number MFCD00084450
InChI Key PPBRXRYQALVLMV-UHFFFAOYSA-N
Synonym ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
PubChem CID 7501
ChEBI CHEBI:27452
SMILES *-CC(-*)C1=CC=CC=C1

Specifications

Physical Form Pellets
Melting Point 93.0°C
Density 1.0500g/mL
Quantity 25 g
Infrared Spectrum Authentic
Loss on Drying 1% max.
Packaging Glass bottle
Linear Formula [-CH2CH(C6H5)-]n
Refractive Index 1.5916
Specific Gravity 1.05
Merck Index 14, 8860
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethylbenzene, methyl isobutyl ketone,, thf, benzene, toluene, pyridine, methylenechloride
IUPAC Name ethenylbenzene
Chemical Name or Material Poly(styrene)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.