missing translation for 'onlineSavingsMsg'
Learn More

Poly(styrene sulfonic acid) sodium salt, M.W. 75,000, Thermo Scientific™

Product Code. 15467105
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15467105 50 g 50g
15477105 250 g 250g
15457105 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15467105 Supplier Thermo Scientific Alfa Aesar Supplier No. 045851.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25704-18-1
Molecular Formula (C8H7NaO3S)n
Molecular Weight (g/mol) 184.21
MDL Number MFCD00084449
InChI Key MAGFQRLKWCCTQJ-UHFFFAOYSA-N
Synonym 4-styrenesulfonic acid sodium salt
PubChem CID 131675404
SMILES [Na]OS(=O)(=O)C1=CC=C(C=C1)C(-*)C-*

Specifications

Color Brown
Physical Form Powder
Quantity 50 g
IUPAC Name 4-ethenylbenzene-1-sulfonic acid
Chemical Name or Material Poly (styrene sulfonic acid) sodium salt, M.W. 75,000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.