missing translation for 'onlineSavingsMsg'
Learn More

Poly(vinyl acetate), approx. M.W. 100,000

Product Code. 10523361 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10523361 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Code produit 10523361 Fournisseur Thermo Scientific Acros Code fournisseur 183265000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 9003-20-7 | (C4H6O2)n | 86.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Spécification

CAS 9003-20-7
Physical Form Beads
Molecular Formula (C4H6O2)n
Linear Formula [-CH2CH(O2CCH3)-]n
MDL Number MFCD00084457
Density 1.1800g/mL
Synonym vinyl acetate, acetic acid ethenyl ester, acetic acid vinyl ester, ethenyl ethanoate, 1-acetoxyethylene, vinyl ethanoate, acetoxyethene, vinylacetat, vinyl acetate monomer, vinyl a monomer
Quantity 500 g
InChI Key XTXRWKRVRITETP-UHFFFAOYSA-N
SMILES CC(=O)OC(-*)C-*
IUPAC Name ethenyl acetate
Molecular Weight (g/mol) 86.09
PubChem CID 7904
ChEBI CHEBI:46916
Infrared Spectrum Authentic
Loss on Drying 5% max.
Packaging Glass bottle
Chemical Name or Material Poly(vinyl acetate), Average M.W. 101600
Afficher plus Afficher moins

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.