missing translation for 'onlineSavingsMsg'
Learn More

Poly(vinyl alcohol), 87.0-89.0% hydrolyzed, M.W. approx. 13,000-23,000

Product Code. 15284208 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 kg
Unit Size:
2.5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15284208 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15284208 Supplier Thermo Scientific Acros Supplier No. 180300025
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 9002-89-5 | (C2H4O)n | 44.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 9002-89-5
Melting Point 230.0°C to 240.0°C
pH 4.5 to 6.5 (4% aq. soln.)
Infrared Spectrum Authentic
Molecular Formula (C2H4O)n
Linear Formula [-CH2CH(OH)-]n
MDL Number MFCD00081922
Quantity 2.5 kg
Merck Index 15,7706
Synonym vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval
Solubility Information Solubility in water: soluble in hot water.
InChI Key IMROMDMJAWUWLK-UHFFFAOYSA-N
SMILES OC(-*)C-*
IUPAC Name ethenol
Molecular Weight (g/mol) 44.05
PubChem CID 11199
Viscosity 3.0-3.7 mPa.s (20°C 4% aq.sol.)
Percent Purity 87.0-89%
Chemical Name or Material Poly(vinyl alcohol)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.