missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(vinyl alcohol), 88% hydrolyzed, average M.W. 145,000-180,000
Shop All Thermo Scientific Chemicals ProductsDescription
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tekniske data
| Melting Point | 230.0°C to 240.0°C |
| Solubility Information | Solubility in water: soluble in hot water. |
| Quantity | 500 g |
| Packaging | Plastic bottle |
| Chemical Name or Material | Poly(vinyl alcohol) |
RUO – Research Use Only
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?