missing translation for 'onlineSavingsMsg'
Learn More

Poly(vinyl alcohol), 95% hydrolyzed, average M.W. 95000, ACROS Organics™

Product Code. 10183910 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10183910 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10183910 Supplier Thermo Scientific Acros Supplier No. 183290250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Indicator
TRUSTED_SUSTAINABILITY

Specifications

Color White to Cream
CAS 9002-89-5
Physical Form Granular Powder
Ash 0.9% max.
Molecular Formula (C2H4O)n
Linear Formula [-CH2CH(OH)-]n
MDL Number MFCD00081922
Merck Index 15, 7706
Synonym vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval
Quantity 25 g
InChI Key IMROMDMJAWUWLK-UHFFFAOYSA-N
SMILES OC(-*)C-*
IUPAC Name ethenol
Molecular Weight (g/mol) 44.05
PubChem CID 11199
Percent Purity 95.5 to 96.5% (degree of hydrolysis)
Infrared Spectrum Authentic
Loss on Drying 5% max.
Packaging Glass bottle
Viscosity 27 to 31 mPa·s (20°C, 4% aq. sol.)
Chemical Name or Material Poly(vinyl alcohol), 95% Hydrolyzed, Average M.W. 95000
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.