missing translation for 'onlineSavingsMsg'
Learn More

Poly(vinyl alcohol), 88% hydrolyzed, average M.W. 20000-30000

Product Code. 10083963 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
2.5kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10083963 25 g Plastic bottle 25g
10711244 500 g Plastic bottle 500g
10276983 1 kg Plastic bottle 1kg
11966971 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10083963 Supplier Thermo Scientific Acros Supplier No. 396760250
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 9002-89-5 | (C2H4O)n | 44.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-89-5
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 44.05
MDL Number MFCD00081922
InChI Key IMROMDMJAWUWLK-UHFFFAOYSA-N
Synonym vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval
PubChem CID 11199
IUPAC Name ethenol
SMILES OC(-*)C-*

Specifications

Color White to Yellow
pH 4.5 to 6.5 (4% aq. soln.)
Physical Form Powder
Melting Point 160.0°C to 240.0°C
Ash 90%% (Na2O) max.
Linear Formula [-CH2CH(OH)-]n
Solubility Information Solubility in water: soluble in hot water.
Quantity 25 g
Percent Purity 87.0 to 89.0% (degree of hydrolysis)
Infrared Spectrum Authentic
Packaging Plastic bottle
Viscosity 3.5 to 4.5cP (4% aq. solution, 20°C) Brookfield
Chemical Name or Material Poly(vinyl alcohol)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.