missing translation for 'onlineSavingsMsg'
Learn More

Polyacrylonitrile, MP Biomedicals

Product Code. 11478195 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11478195 5 g 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11478195 Supplier MP Biomedicals Supplier No. 0521753805
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifikationer

CAS 25014-41-9
Molecular Formula C3H3N
Synonym acrylonitrile, 2-propenenitrile, vinyl cyanide, propenenitrile, cyanoethylene, carbacryl, fumigrain, acritet, acrylon, ventox
Quantity 5 g
InChI Key NLHHRLWOUZZQLW-UHFFFAOYSA-N
SMILES C=CC#N
IUPAC Name prop-2-enenitrile
Molecular Weight (g/mol) 53.064
PubChem CID 7855
ChEBI CHEBI:28217
Chemical Name or Material Polyacrylonitrile
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.