missing translation for 'onlineSavingsMsg'
Learn More

Polyacrylonitrile, TRC

Product Code. 30426725
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30426725 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30426725 Supplier Toronto Research Chemicals Supplier No. TRCP68815310G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Polyacrylonitrile
CAS 25014-41-9
Molecular Formula (C3H3N)x
Quantity 10 g
Synonym ACL 1050, ACP 0013, ACP 1010, Acrilan 16
InChI Formula InChI=1S/C3H3N/c1-2-3-4/h2H,1H2
SMILES C=CC#N
Molecular Weight (g/mol) 53.06
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.