Learn More
Nickel Electro™ Polycarbonate Hinged Gable Lid
Product Code. 12758888 Shop All Nickel-Electro Products
Click to view available options
For Use With (Equipment):
22 and 28 L Unstirred/Shaking Baths
8 and 10 L Unstirred/Shaking Baths
Unit Size:
Each
Product Code. 12758888
Brand: Nickel Electro™ GL18
Only null left
Add to basket
Add to basket
Maximize bath area with a hinged lid that drips condensation back into bath. Nickel Electro™ Polycarbonate Hinged Gable Lid are transparent to allows samples to be viewed.
  • Lightweight
  • Semi-transparent for viewing samples
  • Maximize bath area
Specifications
For Use With (Equipment) 8 and 10 L Unstirred/Shaking Baths
For Use With (Application) Reducing Water Evaporation, Keeping Samples and Water Free From Contamination and Saves Energy
Material Polycarbonate