Learn More
Masterflex™ Polycarbonate Luer Fittings
Product Code. 11760279
Change view
Click to view available options
For Use With:
Female Luer x 1/4-28 UNF Thread
Female Luer x 1/8 in. Hose Barb
Female Luer x 10-32 in. UNF Thread
Male Luer w/ Lock Ring x 1/16 in. Hose Barb
Male Luer w/ Lock Ring x 1/4 in. Hose Barb
Male Luer w/ Lock Ring x 1/8 in. Hose Barb
Male Luer x 10-32 in. Thread
Male Luer x Female Luer Coupler
Male luer x male luer adapter
Unit Size:
Pack of 25
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With unitSize
11760279 Female Luer x 1/4-28 UNF Thread Pack of 25
11746008 Female Luer x 10-32 in. UNF Thread Pack of 25
11745988 Male Luer x 10-32 in. Thread Pack of 25
11749959 Female Luer x 1/8 in. Hose Barb Pack of 25
12197750 Male Luer w/ Lock Ring x 1/16 in. Hose Barb Pack of 25
11739709 Male Luer w/ Lock Ring x 1/4 in. Hose Barb Pack of 25
11729709 Male Luer w/ Lock Ring x 1/8 in. Hose Barb Pack of 25
11745668 Male Luer x Female Luer Coupler Pack of 25
11959668 Male luer x male luer adapter Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 11760279 Supplier Masterflex™ Supplier No. 4550166
Only null left
Add to basket
Add to basket
Designed to work with microtubing and luer-type syringes. Standard-sized connecting ends make these luer fittings completely interchangeable for surefire installations.
  • Manufactured from 100% virgin raw materials
  • Free of contaminants and ideal for syringes or small tubing
  • Choose from a range of sizes with a variety of connection options
  • Easily connect fluid paths without the use of tools
  • Male and female luers available
Specifications
Certifications/Compliance ISO 9001
Material Polycarbonate
For Use With Female Luer x 1/4-28 UNF Thread
For Use With (Application) Transitions between luer fittings and UNF or NPT threads
Packaging Quantity 25 Pack
Thread Size 1/4 to 28 UNF
Product Type Luer Fitting