missing translation for 'onlineSavingsMsg'
Learn More

Polydialdehyde Starch (Polymeric Dialdehyde), TRC

Product Code. 30135588
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135588 5 g 5g
30012471 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135588 Supplier Toronto Research Chemicals Supplier No. TRCP6880635G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9047-50-1
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.22
SMILES CC(C=O)C(CC(C=O)OC)CO

Specifications

Quantity 5 g
InChI Formula InChI=1S/C9H16O4/c1-7(4-10)8(5-11)3-9(6-12)13-2/h4,6-9,11H,3,5H2,1-2H3
Chemical Name or Material Polydialdehyde Starch (Polymeric Dialdehyde)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.