missing translation for 'onlineSavingsMsg'
Learn More

Polydimethylsiloxane (silicone) emulsion

Product Code. 15462147
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15462147 500 g 500g
15452147 100 g 100g
15472147 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15462147 Supplier Thermo Scientific Alfa Aesar Supplier No. 045088.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Polydimethylsiloxane (silicone) emulsion is the most widely used silicon-based organic polymer and is particularly known for its unusual rheological (or flow) properties. Further, it is used in contact lenses, medical devices to elastomers and in shampoos (as dimethicone makes hair shiny and slippery). It finds application as an antifoaming agent in food, caulking, lubricants, kinetic sand and heat-resistant tiles. In addition to this, it serves as a critical ingredient in water-repelling coatings such as Rain-X.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00132673

Specifications

Physical Form Liquid
Solubility Information Miscible with carbon tetrachloride,benzene,chloroform,diethyl ether,toluene and other organic solvents. Immiscible with water and ethanol.
Quantity 500 g
Chemical Name or Material Polydimethylsiloxane (silicone) emulsion

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.