missing translation for 'onlineSavingsMsg'
Learn More

Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 139,000

Product Code. 11465470
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
4 kg
Unit Size:
1kg
250g
4kg
50g
Product Code. Quantity unitSize
11465470 50 g 50g
11475470 250 g 250g
11485470 1 kg 1kg
11495470 4 kg 4kg
4 options
This item is not returnable. View return policy

Product Code. 11465470

Brand: Thermo Scientific Alfa Aesar 044644.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9016-00-6 | (C2H6OSi)n | 74.15 g/mol

Polydimethylsiloxane, trimethylsiloxy is used in water-repellent, protective creams & lotions. It is also used as surfactant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9016-00-6
Molecular Formula (C2H6OSi)n
Molecular Weight (g/mol) 74.15
MDL Number MFCD00084411
InChI Key SEUDSDUUJXTXSV-UHFFFAOYSA-N
IUPAC Name dimethylsilanone
SMILES C[Si](C)(-*)O-*

Specifications

Color Colorless
Physical Form Viscous Liquid
Boiling Point >205°C
Melting Point <-40°C
Density 0.977 g/mL
Solubility Information Not miscible with water. Insoluble in methanol,and ethylene glycol.
Quantity 50 g
Flash Point 321°C (610°F)
Odor Odorless
Refractive Index 1.4035
Chemical Name or Material Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 139,000
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.