missing translation for 'onlineSavingsMsg'
Learn More

Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 237

Product Code. 11379028
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
Unit Size:
1 L
250mL
50mL
Product Code. Quantity unitSize
11379028 50 mL 50mL
11389028 250 mL 250mL
11399028 1 L 1 L
3 options
This item is not returnable. View return policy

Product Code. 11379028

Brand: Thermo Scientific Alfa Aesar 042496.AD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9016-00-6 | (C2H6OSi)n | 74.15 g/mol

Polydimethylsiloxane, trimethylsiloxy is used in water-repellent, protective creams & lotions. It is also used as surfactant, is used in coating auxiliary agents. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9016-00-6
Molecular Formula (C2H6OSi)n
Molecular Weight (g/mol) 74.15
MDL Number MFCD00084411
InChI Key SEUDSDUUJXTXSV-UHFFFAOYSA-N
Synonym Hexamethyldisiloxane polymers
IUPAC Name dimethylsilanone
SMILES C[Si](C)(-*)O-*

Specifications

Color Colorless
Physical Form Liquid
Boiling Point 152°C to 153°C
Melting Point -86°C
UN Number UN1993
Density 0.820 g/mL
Solubility Information Soluble in aromatic hydrocarbon,partially soluble in aliphatic and insoluble in water.
Quantity 50 mL
Flash Point 39°C (102°F)
Odor Odorless
Refractive Index 1.3825
Chemical Name or Material Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 237
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.