missing translation for 'onlineSavingsMsg'
Learn More

Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 4000

Product Code. 11399038
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
Unit Size:
1 L
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11399038 50 mL 50mL
11319048 250 mL 250mL
11309048 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11399038 Supplier Thermo Scientific Alfa Aesar Supplier No. 042500.AD
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 9016-00-6 | (C2H6OSi)n | 74.15 g/mol

Polydimethylsiloxane, trimethylsiloxy terminated is widely used as a common surfactant. It is an active component of defoamers which suppress the formation of foams and as a critical ingredient in water-repelling coatings as an active silicone fluid. It finds application in optic telecommunications, biomedical research, elasticity of polymer networks, adhesive materials and in skin-moisturizing lotions. Furthermore, it is used as a filler fluid in breast implants and as a sorbent for the analysis of headspace (dissolved gas analysis) of food.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9016-00-6
Molecular Formula (C2H6OSi)n
Molecular Weight (g/mol) 74.15
MDL Number MFCD00084411
InChI Key SEUDSDUUJXTXSV-UHFFFAOYSA-N
IUPAC Name dimethylsilanone
SMILES C[Si](C)(-*)O-*

Specifications

Color Colorless
Physical Form Viscous Liquid
Boiling Point >225°C
Melting Point <-40°C
Density 0.960 g/mL
Solubility Information Slightly miscible with water. Immiscible with methanol and ethylene glycol.
Quantity 50 mL
Flash Point 285°C (545°F)
Odor Odorless
Refractive Index 1.4015
Chemical Name or Material Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 4,000
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.