missing translation for 'onlineSavingsMsg'
Learn More

Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 9000

Product Code. 11369048
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
Unit Size:
1 L
250mL
50mL
Product Code. Quantity unitSize
11369048 50 mL 50mL
11379048 250 mL 250mL
11359048 1 L 1 L
3 options
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 11369048

Brand: Thermo Scientific Alfa Aesar 042502.AD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

CAS: 9016-00-6 | (C2H6OSi)n | 74.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 9016-00-6
Molecular Formula (C2H6OSi)n
Molecular Weight (g/mol) 74.15
MDL Number MFCD00084411
InChI Key SEUDSDUUJXTXSV-UHFFFAOYSA-N
IUPAC Name dimethylsilanone
SMILES C[Si](C)(-*)O-*

Tekniske data

Color Colorless
Physical Form Viscous Liquid
Boiling Point >205°C
Melting Point <-40°C
Density 0.968 g/mL
Quantity 50 mL
Flash Point 315°C (599°F)
Odor Odorless
Refractive Index 1.403
Chemical Name or Material Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 9,000

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.