missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Polyester Cable

Product Code. 11746088 Shop All Cole-Parmer Products
Change view
Click to view available options
Length (Metric):
30 m
60 m
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
11746088 60 m Each
11736088 30 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11746088 Supplier Cole-Parmer™ Supplier No. 0549936
Estimated Shipment: 22-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use this solid braided line with sampling equipment that weighs less than 75 lb (34 kg)

  • Maximum load is 110 lb (50 kg)

Specifications

Product Type Cable
Material Polyester
Quantity 1 Ea.
Length (Metric) 60 m
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.