missing translation for 'onlineSavingsMsg'
Learn More

Bollé Safety Polyester Neck Cord

Product Code. 11803480
Click to view available options
Color:
Black
Blue, green
Unit Size:
Each
Pack of 10
This item is not returnable. View return policy

Product Code. 11803480

Brand: Bollé Safety CORD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Keep track of your Bolle Protection™ safety glasses with the Polyester Neck Cord, a helpful accessory.

  • Polyester
TRUSTED_SUSTAINABILITY

Specifications

Color Black
Product Type Neck Cord
For Use With (Application) Bollé Safety Spectacles
Length (Metric) 600 mm
Model CORD
No. per Pack 1 Ea.
Material Polyester
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.