Learn More
Macherey-Nagel™ Polyester Sheets Polygram SIL G/UV254 TLC Micro-Set F2
Brand: Macherey-Nagel™ 814300
Code : NEW
Additional Details : Weight : 0.26500kg Transport : UN number : 3316 Chem class : 9 Pack group : II
Specifications
Polyester Sheets Polygram SIL G | |
TLC Micro-Set F2 | |
1 manual; 50 glass capillaries 1 μL;50 polyester sheets Polygram SIL G/UV254, 4 x 8 cm; 5 disposable pipettes 25 μL; 5 sample vials N 11, 1.5 mL, with PE caps and seals; 3 sample vials 30 mL (for butter, margarine and edible oil); 100 mL each of cyclohexane and molybdatophosphoric acid spray reagent (in ethanol)*; 2 x 50 mL acetone with calibrated pipette (1 x enclosed)*; 25 mL butan-2-one (ethyl methyl ketone)*;8 mL cholesterol reference solution (in acetone)* |
1 Ea. | |
UV254 | |
4 x 8 cm |
Safety and Handling
- TLC Micro Set F 2
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H336-May cause drowsiness or dizziness.
- H410-Very toxic to aquatic life with long lasting effects.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P331-Do NOT induce vomiting.
Supplemental information
- MIXTURE LIST-Contains : cyclohexane, acetone, ethyl methyl ketone, ethanol, molybdatophosphoric acid