missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Polyester Transport Signs

Brand:  Brady™ 257506

 View more versions of this product

Product Code. 10020244

  • 5.10 € / Each
Only null left
Estimated Shipment: 06-08-2025
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

ISO 7010
Gloss
1 Ea.
Pipemarkers and safety signs
0.024 mm
-40 °C-110 °C
Flammable Solid 4
Red on White
ADR 4.1-100*100-B7541
Polyester
Permanent Acrylic
100 mm
100 mm
Diamond
Transport Sign
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.