Specifications
| Material | PEEK, Silica |
| Type | Microfraction Collection Kit |
| Includes | 1 x Tubing with one sleeve preassembled, i.d. 0.15 mm, L=800 mm (Silica), 1 x Sleeve o.d. 1/16 in., i.d. 405 μm (Silica), 2 x Finger tight narrow, head connector (PEEK), for 1/16 in. o.d. tubing, 1 x Instruction for Micro fraction collection |
| Quantity | 1 Kit |