missing translation for 'onlineSavingsMsg'
Learn More

Cytiva PEEK Sample Loops

Product Code. 11370032 Shop All Cytiva Products
Change view
Click to view available options
Volume (Metric):
100 μL
500 μL
1 mL
2 mL
5 mL
Diameter (Metric) Inner:
0.5 mm
0.75 mm
1 mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Diameter (Metric) Inner Unit Size
11370032 1 mL 0.75 mm Each
11360032 100 μL 0.5 mm Each
11390032 2 mL 0.75 mm Each
10852041 5 mL 1 mm Each
11306971 500 μL 0.75 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11370032 Supplier Cytiva Supplier No. 18111401
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Resists organic solvents and salt buffers commonly used in chromatography of biomolecules. Cytiva Life Sciences™ PEEK Sample Loops are an important accessory for chromatography applications.

  • Wetted parts are resistant to organic solvents
  • Wetted parts are resistant to salt buffers
  • Except 100 % Ethyl acetate, 100 % Hexane and 100 % Tetrahydrofuran
  • Polyetheretherketone (PEEK)
TRUSTED_SUSTAINABILITY

Specifications

Type Sample Loop
Material PEEK
Connection Size (Metric) 1.58 mm
Diameter (Metric) Inner 0.75 mm
Volume (Metric) 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.