missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Polyethersulfone Membranes

Product Code. 11785305 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Pore Size:
0.2 μm
0.45 μm
Diameter (Metric):
25 mm
47 mm
Unit Size:
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Diameter (Metric) Unit Size
11785305 0.45 μm 47 mm Pack of 100
11755305 0.2 μm 25 mm Pack of 100
11765305 0.2 μm 47 mm Pack of 100
11775305 0.45 μm 25 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11785305
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

Fisherbrand™ Polyethersulfone Membranes features highly asymmetrical pore structure that offers effective loading capacity and high flow rate.

  • Highly asymmetrical pore structure offers effective loading capacity and high flow rate
  • Hydrophilic and made with polyethersulfone polymer
  • Designed to remove particulates during general filtration
  • Low protein and drug binding characteristics make it ideally suited for use in life science applications

Specifications

Autoclavable Yes
Bubble Point 2.7bar
Color White
Material PES
Pore Size 0.45 μm
Sterility Non-sterile
Flow Rate 21mL/min./cm2
For Use With (Application) Filtration of special chemical reagent, high temperature liquids, filtration of special chemical reagent, high temperature liquids, filtration of drugs
Gas Flow Rate 16L/min./cm2
Hydrophobicity Hydrophilic
Quantity 100 Pack
Type Plain Disc
Wettability Yes
Diameter (Metric) 47 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.