missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Included in the ÄKTAdesign XT Kit for connecting Autosampler A-900 or A-905 to the ÄKTAexplorer system.
Specifications
Specifications
| Type | Union |
| Connections | M6 Female, 1/16 in. |
| Material | PEEK |
| Color | Black |
| For Use With (Equipment) | ÄKTA Chromatography Systems, A-900 or A-905 Autosampler |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?