missing translation for 'onlineSavingsMsg'
Learn More

Sperian™ Polyethylene Coveralls

Product Code. p-8010197
Click to view available options
Size:
Medium
Large
Unit Size:
Pack of 25
This item is not returnable. View return policy

Product Code. 11856910

Brand: Sperian™ 4571433M

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Product Type Single-use Coveralls
Size Medium
Gender Unisex
Material Polypropylene
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Hood Type Elastic
Sterility Non-sterile
Resistance Tears
Certifications/Compliance (Europe) EC Category PPE, Cat III to Directive 89/686/EEC
For Use With (Application) Chemical Industries, Maintenance, Medical and Pharmaceuticals, Spray Painting, Automotive
Weight (Metric) 55 g/m2
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.