missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Polyethylene glycol 12-hydroxystearate

Product Code. 16465467
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16465467 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16465467 Supplier MedChemExpress Supplier No. HYY1893

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Polyethylene glycol 12-hydroxystearate, a Macrogol 15 hydroxy stearate, is a permeability enhancer.

Specifications

CAS 61909-81-7
Solubility Ethanol : 100 mg/mL (Need ultrasonic)
Content And Storage 4°C, sealed storage, away from moisture
Quantity 50 g
Product Type Biochemical Assay Reagents
Shipping Condition Room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.