Learn More
Description
- Differential precipitation of DNA.
- Enhances hybridization rate of nucleic acids.
- Improves efficiency of end-labeling with T4 Polynucleotide Kinase(1,2).
- Used in ligation of blunt ended DNA.
- Precipitates bacteriophage ± particles and plasmid DNA.
- Used in preparing single-stranded M13 DNA.
- Promotes cell fusion.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Differential precipitation of DNA
- Enhances hybridization rate of nucleic acids
- Improves efficiency of end-labeling with T4 Polynucleotide Kinase(1,2)
- Used in ligation of blunt ended DNA
- Precipitates bacteriophage &
Specifications
Specifications
| Physical Form | Flakes |
| Melting Point | 60°C to 63°C |
| Quantity | 1 kg |
| Packaging | Plastic bottle |
| Chemical Name or Material | Polyethylene Glycol 6000 |
RUO – Research Use Only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.