missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene Glycol 8000 (PEG), Fisher BioReagents™

Product Code. 10224963
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Glass Bottle
Poly Bottle
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10224963 100 g Glass Bottle 100g
10407773 1 kg Poly Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10224963 Supplier Fisher BioReagents Supplier No. BP233100
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Polyethylene Glycol 8000, HO-[CH2CH2O]n-H, CAS Number-25322-68-3, Polyethylene Oxide, Carbowax, PEG 8000, 1kg, Gray, White, 7000 to 9000, CAUTION!

PEG 8000 is suitable as a medium for the fusion of mammalian cells. It is also used to precipitate bacteriophage from lysed cell supernatants.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym PEG 8000, Polyethylene Oxide, Carbowax™
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

Color White
pH 5.0 to 7.0
Physical Form Solid
Melting Point 55°C
Linear Formula H(OCH2CH2)nOH
Merck Index 15, 7688
Quantity 100 g
Formula Weight 7000 to 9000
Identification Pass Test
Packaging Glass Bottle
Chemical Name or Material Polyethylene Glycol 8000
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.