missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene glycol diacrylate, M.W. 1,000

Product Code. 15402707
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15402707 250 mg 250mg
15482697 1 g 1g
15492697 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15402707 Supplier Thermo Scientific Alfa Aesar Supplier No. 046493.MD
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 26570-48-9 | C5H10O4 | 134.131 g/mol

PEGDA is used in dental compounds, offset inks, varnishes and flexible coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26570-48-9
Molecular Formula C5H10O4
Molecular Weight (g/mol) 134.131
MDL Number MFCD00081876
InChI Key FCDZZFLRLVQGEH-UHFFFAOYSA-N
Synonym acrylic acid; ethylene glycol, poly ethylene glycol diacrylate, polyethylene glycol diacrylate, poly ethylene glycol acrylate, average mn ∼375
PubChem CID 16212859
IUPAC Name ethane-1,2-diol;prop-2-enoic acid
SMILES C=CC(=O)O.C(CO)O

Specifications

Color White
Physical Form Granular Solid
Solubility Information Soluble in THF and chloroform.
Quantity 250 mg
Sensitivity Light sensitive
Chemical Name or Material Polyethylene glycol diacrylate, M.W. 1,000
compliance-icons compliance-icons
Product Identifier
  • Polyethylene glycol diacrylate, M.W. 1,000
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Polyethylene glycoldiacrylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.