missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene Glycol Diamine (MW 20,000), TRC

Product Code. 30417360
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30417360 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30417360 Supplier Toronto Research Chemicals Supplier No. TRCP688503250MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Polyethylene Glycol Diamine
CAS 24991-53-5
Molecular Formula (C2H4O)nC4H12N2O
Quantity 250 mg
Synonym Diamine-PEG 897, Diamino-terminated Polyethylene Glycol, Fluka 06703, JSJ 73-006, Jeffamine 2001, O,O'-Bis(2-aminoethyl)octadecaethylene Glycol, PEG 1000 Diamine, PEG 4000DCA, PEG Diamine, PEO 600, PEO-Amine 400, Poly(ethylene oxide)diamine, Poly(oxyethylene)diamine, Polyethylene Glycol Bis(2-aminoethyl) Ether, Tetradecaethylene Glycol Diamine, Y 10000, Y 20000, Y 4000
InChI Formula InChI=1S/C6H16N2O2/c7-1-3-9-5-6-10-4-2-8/h1-8H2
SMILES NCCOCCOCCN
Molecular Weight (g/mol) 148.2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.