missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene Glycol Glycidyl Lauryl Ether, TRC

Product Code. 30417962
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30417962 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30417962 Supplier Toronto Research Chemicals Supplier No. TRCP688518500MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Polyethylene Glycol Glycidyl Lauryl Ether
CAS 86630-59-3
Molecular Formula (C2H4O)n . C15H30O2
Quantity 500 mg
Synonym Poly(oxy-1,2-ethanediyl)','α-dodecyl-ω-(oxiranylmethoxy)- (9CI), Denacol EX 171, Denacol FCA 050, Emulgen 120GE, EX 171, Lauryl (EO)15 glycidyl ether, Polyethylene glycol dodecyl glycidyl ether, Polyethylene glycol glycidyl lauryl ether, Polyethylene glycol lauryl glycidyl ether, Polyethylene glycol monododecyl monoglycidyl ether, Polyethylene glycol monoglycidyl monolauryl ether, Polyoxyethylene glycidyl lauryl ether
InChI Formula InChI=1S/C17H34O3/c1-2-3-4-5-6-7-8-9-10-11-12-18-13-14-19-15-17-16-20-17/h17H,2-16H2,1H3
SMILES CCCCCCCCCCCCOCCOCC1CO1
Molecular Weight (g/mol) 286.4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.