missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene Glycol, MP Biomedicals™

Product Code. 11487152 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11487152 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11487152 Supplier MP Biomedicals Supplier No. 02151916.5
  • ONLINE EXCLUSIVE PRICE
    115.00 €
    108.15 € / 500g
    Save 6.85 €
    5% Off
 Restricted Item
Estimated Shipment: 29-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Formed as a condensation polymer of ethylene oxide and water. Polyethylene Glycol, MP Biomedicals (PEG) has been used in many applications from medicine to industrial manufacturing, including the precipitation of proteins.

PEG has been used in a wide variety of biomedical and laboratory applications, including:

  • The precipitation of proteins
  • As a fusing agent in enhancing the effect of macrophages on hybridoma
  • In the separation and purification of biomolecules
  • In induction of cell hybridization
  • To recover pathogenic viruses and human rotaviruses from water, oyster and sediment samples
  • Demonstrated to have anticell proliferation and antitumor effects via induction of apoptosis

Utilization of PEG:

  • PEGs are susceptible to oxidative degradation in the presence of air; minimizing exposure to elevated temperatures and oxygen or addition of an antioxidant can limit the amount of degradation
  • PEGs do not hydrolyze or deteriorate upon storage
  • PEGs do not support the growth of molds
  • PEG is incompatible with phenol and may reduce the antimicrobial action of other preservatives

Specifications

CAS 107-21-1
Melting Point 55°C to 65°C
Density 1.1–1.2g/mL (Lit.)
pH 4.5 to 7.5 (5% aq. soln.)
Molecular Formula C2H6O2
Quantity 500 g
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, polyethylene glycol, macrogol
Solubility Information Soluble in aqueous solution (100mg/mL, clear and colorless solution).
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Molecular Weight (g/mol) 62.07
PubChem CID 174
ChEBI CHEBI:30742
Formula Weight 3,400 average
Vapor Pressure <0.01mmHg at 20°C (Lit.)
Percent Purity 97 to 103%
Grade Not Available
Physical Form Powder
Chemical Name or Material Polyethylene Glycol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.