missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene (High Density), TRC

Product Code. p-200172747
Change view
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006550 100 g 100g
30004820 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30006550 Supplier Toronto Research Chemicals Supplier No. TRCP688285100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-88-4
Molecular Formula C2 H4
Molecular Weight (g/mol) 28.053
Synonym Polyethylene, Poly(ethylene), Ethene, homopolymer, Ethylene, polymers (8CI);
SMILES CCCC

Specifications

Quantity 100 g
InChI Formula InChI=1S/C2H4/c1-2/h1-2H2
Formula Weight 28.031
Chemical Name or Material Polyethylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.