missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene, low density, 1000 micron

Product Code. 11354998
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
Product Code. Quantity unitSize
11354998 50 g 50g
11334998 250 g 250g
11344998 1 kg 1kg
3 options
This item is not returnable. View return policy

Product Code. 11354998

Brand: Thermo Scientific Alfa Aesar 043949.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9002-88-4 | (C2H4)n | 28.05 g/mol

Polyethylene is used as an additive to unsaturated polyesters, epoxides and other polymers to impart the unique properties of ultra-high-molecular-weight polyethylene (UHMWPE). It is widely used for the manufacturing of various containers, trays, dispensing bottles, plastic wraps, plastic bags for computer components and various molded laboratory equipments. It is used in the manufacture of plastic bags, yard bags or outdoor bags, which is an environmentally sensible material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-88-4
Molecular Formula (C2H4)n
Molecular Weight (g/mol) 28.05
MDL Number MFCD00084423
InChI Key VGGSQFUCUMXWEO-UHFFFAOYSA-N
Synonym ethylene, acetene, elayl, olefiant gas, athylen, etileno, liquid ethylene, polyethylene, bicarburretted hydrogen, plastipore
PubChem CID 6325
ChEBI CHEBI:18153
IUPAC Name ethene
SMILES *-CC-*

Specifications

Physical Form Granular
Merck Index 14,7567
Solubility Information Insoluble in water.
Quantity 50 g
Odor Odorless
Chemical Name or Material Polyethylene, Low density, 1000 micron

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.