missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene, low density, ∣<400 micron

Product Code. 11369228
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
2500 g
Unit Size:
100g
2500g
25g
500g
5g
Product Code. Quantity unitSize
11369228 5 g 5g
11349228 25 g 25g
11379228 100 g 100g
11359228 500 g 500g
11366119 2500 g 2500g
5 options
This item is not returnable. View return policy

Product Code. 11369228

Brand: Thermo Scientific Alfa Aesar 042607.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9002-88-4 | (C2H4)n | 28.05 g/mol

Polyethylene is used in injection and blow molded toys, housewares, lids, injection molded car seats, Hot melt coating for paper, additive in cast moldings, candles, oil-based inks and hot melt adhesives. It is also used as an additive, lubricant in rubber processing, extrusion and calendaring aid for PVC and dispersing aid for color concentrates. Bearings, gears, bushings and other moving parts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-88-4
Molecular Formula (C2H4)n
Molecular Weight (g/mol) 28.05
MDL Number MFCD00084423
InChI Key VGGSQFUCUMXWEO-UHFFFAOYSA-N
Synonym ethylene, acetene, elayl, olefiant gas, athylen, etileno, liquid ethylene, polyethylene, bicarburretted hydrogen, plastipore
PubChem CID 6325
ChEBI CHEBI:18153
IUPAC Name ethene
SMILES *-CC-*

Specifications

Physical Form Powder
Density 0.92 g/mL
Merck Index 14,7567
Solubility Information Insoluble in water.
Quantity 5 g
Odor Odorless
Chemical Name or Material Polyethylene, Low density, ≤400 micron

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.