Learn More
Fisherbrand™ Polyethylene Overlay Sheet
Product Code. 12378546
Change view
Click to view available options
For Use With (Equipment):
Fisherbrand GD-4534 Gel Dryer System
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
12378546 Fisherbrand GD-4534 Gel Dryer System Each
1 options
Product Code. 12378546 Supplier Fisherbrand™ Supplier No. GDPPS
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Accessory for use with vacuum gel dryers.
  • Detachable
  • Allows gel dryer to be adapted for thin polyacrylamide gel, thicker agarose gels
  • Replacement part for Vacuum Gel Dryer systems
  • Pore size of porous polyethylene cover sheet:  0.75mm
Specifications
For Use With (Equipment) Fisherbrand GD-4534 Gel Dryer System
Type Sheet