missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene oxide, M.W. 300,000
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
Description
Polyethylene oxide, M.W. 300,000 is used to impart viscosity to and modify flow of aqueous solutions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| pH | 5 to 7 |
| Physical Form | Powder |
| Melting Point | 65°C |
| Density | 1.21 g/mL |
| Solubility Information | Soluble in acetone,alcohol,chloroform,toluene and dichloromethane. |
| Quantity | 500 g |
| Odor | Odorless |
| Refractive Index | 1.4539 |
| Chemical Name or Material | Polyethylene oxide, M.W. 300,000 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction