missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene rod, High Density, 6.35mm (0.25 in.) dia., Thermo Scientific™

Product Code. 15476785
Change view
Click to view available options
Quantity:
30 cm
Unit Size:
30cm
Product Code. Quantity unitSize
15476785 30 cm 30cm
1 options
This item is not returnable. View return policy

Product Code. 15476785

Brand: Thermo Scientific Alfa Aesar 045159.BV

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 9002-88-4
Physical Form Rod
Molecular Formula (C2H4)n
MDL Number MFCD00084423
Synonym ethylene, acetene, elayl, olefiant gas, athylen, etileno, liquid ethylene, polyethylene, bicarburretted hydrogen, plastipore
Quantity 30 cm
InChI Key VGGSQFUCUMXWEO-UHFFFAOYSA-N
SMILES *-CC-*
IUPAC Name ethene
Molecular Weight (g/mol) 28.05
PubChem CID 6325
ChEBI CHEBI:18153
Odor Odorless
Chemical Name or Material Polyethylene rod, High Density, 6.35mm (0.25 in.) dia.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.