missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene rod, Low Density, 19mm (0.75 in.) dia., Thermo Scientific™

Product Code. 15416835
Change view
Click to view available options
Quantity:
30 cm
Unit Size:
30cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15416835 30 cm 30cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15416835 Supplier Thermo Scientific Alfa Aesar Supplier No. 045219.BV
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 9002-88-4
Physical Form Rod
Molecular Formula (C2H4)n
MDL Number MFCD00084423
Synonym ethylene, acetene, elayl, olefiant gas, athylen, etileno, liquid ethylene, polyethylene, bicarburretted hydrogen, plastipore
Quantity 30 cm
InChI Key VGGSQFUCUMXWEO-UHFFFAOYSA-N
SMILES *-CC-*
IUPAC Name ethene
Molecular Weight (g/mol) 28.05
PubChem CID 6325
ChEBI CHEBI:18153
Odor Odorless
Chemical Name or Material Polyethylene rod, Low Density, 19mm (0.75 in.) dia.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.