Learn More
Fisherbrand™ Polyethylene Sample Bags
For use in the laboratory to carry samples into a sterile environment.
Brand:  Fisherbrand™ 14955192
Product Code. 11994385
  • ONLINE EXCLUSIVE PRICE
    93.80 €
    42.40 € / Pack of 250
    Save 51.40 €
    54% Off
Only null left
Estimated Shipment: 09-06-2026
Add to basket
Description
Description
  • Secure, contaminant free bags to ensure dependable analysis results
  • Made from heavy gauge, virgin polyethylene that meets FDA specifications
  • Can accommodate solid, semi solid and liquid materials
  • Easy to use, insert sample and seal by twirling the bag away outward four times and bending wire ends inward
  • Economical and efficient way to collect, contain and carry samples