missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene sheet, Low Density, 1.6mm (0.063in) thick

Product Code. 15446785
Change view
Click to view available options
Quantity:
300 x 300 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15446785 300 x 300 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15446785 Supplier Thermo Scientific Alfa Aesar Supplier No. 045154.HB

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 9002-88-4 | (C2H4)n | 28.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Color White
CAS 9002-88-4
Molecular Formula (C2H4)n
MDL Number MFCD00084423
Synonym ethylene, acetene, elayl, olefiant gas, athylen, etileno, liquid ethylene, polyethylene, bicarburretted hydrogen, plastipore
Quantity 300 x 300 mm
InChI Key VGGSQFUCUMXWEO-UHFFFAOYSA-N
SMILES *-CC-*
IUPAC Name ethene
Molecular Weight (g/mol) 28.05
PubChem CID 6325
ChEBI CHEBI:18153
Odor Odorless
Chemical Name or Material Polyethylene sheet, Low Density, 1.6mm (0.063 in.) thick
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.