Learn More
Gravis Polyethylene Snap-Caps
Product Code. 11773883
Click to view available options
Thread Size (Metric):
22 mm
30 mm
40 mm
Unit Size:
Pack of 100
Pack of 86
Product Code. 11773883
Brand: Gravis Q40212
Only null left
Add to basket
Edge
Add to basket
Providing secure closure for glass sample bottles. Gravis Polyethylene Snap-Caps are available for different size closures.
  • Durable
  • Provides excellent chemical resistance
Specifications
Material Polyethylene
No. per Pack 86Pack
Quantity 86 Pack
Thread Size (Metric) 40 mm
Product Type Snap Cap
Closure Type Snap